missing translation for 'onlineSavingsMsg'
Learn More

(-)-Caryophyllene oxide, 95%

Product Code. 10217190 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 10217190

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1139-30-6 | C15H24O | 220.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1139-30-6
Molecular Formula C15H24O
Molecular Weight (g/mol) 220.35
MDL Number MFCD00134216
InChI Key NVEQFIOZRFFVFW-RGCMKSIDSA-N
Synonym caryophyllene oxide, --caryophyllene oxide, beta-caryophyllene epoxide, beta-caryophyllene oxide, --epoxycaryophyllene, unii-s2xu9k448u, -carophyllene oxide, caryophyllene epoxide, epoxycaryophyllene, trans-caryophyllene oxide
PubChem CID 1742210
ChEBI CHEBI:67818
SMILES CC1(CC2C1CCC3(C(O3)CCC2=C)C)C

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 55°C to 62°C
Color White
Density 0.9600g/mL
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4956
Quantity 10 g
Beilstein 17, IV, 392
Specific Gravity 0.96
Specific Rotation -55
Formula Weight 220.35
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material (-)-Caryophyllene oxide
Show More Show Less
compliance-icons
Product Identifier
  • (-)-Caryophyllene oxide
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : (-)-Caryophyllene oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.