missing translation for 'onlineSavingsMsg'
Learn More

Casein Kinase II Inhibitor IV, MedChemExpress

Product Code. 16470747
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16470747 50 mg 50mg
16467157 5 mg 5mg
16435326 100 mg 100mg
16424438 10 mg 10mg
16423086 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16470747 Supplier Medchem Express Supplier No. HY111378
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Casein Kinase II Inhibitor IV is a potent, ATP-competitive of casein kinase II inhibitor with an IC50 of 9 nM. Casein Kinase II Inhibitor IV ia also a human keratinocytes (NHEK) differentiation inducer.

Casein Kinase II Inhibitor IV is a potent, ATP-competitive of casein kinase II inhibitor with an IC50 of 9 nM. Casein Kinase II Inhibitor IV ia also a human keratinocytes (NHEK) differentiation inducer.

Specifications

Chemical Name or Material Casein Kinase II Inhibitor IV
CAS 863598-09-8
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C24H23N5O3
Quantity 50 mg
Solubility Information DMSO : 33.33 mg/mL (77.61 mM; Need ultrasonic)
SMILES N#CCCC1=CC=CC(N2C=CC3=CN=C(NC4=CC(OC)=C(OC)C(OC)=C4)N=C32)=C1
Molecular Weight (g/mol) 429.47
Formula Weight 429.47
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.