missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Casein Kinase II Substrate

Product Code. 15497719
Click to view available options
Substrate Quantity:
0.5mg
1mg
Unit Size:
0.5mg
1mg
This item is not returnable. View return policy

Product Code. 15497719

Brand: Thermo Scientific™ J66383.M^

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For cellular and molecular biology applications

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Color White to off-white
Form Lyophilized
Molecular Weight (g/mol) 1362.38
Storage Requirements -30° to -10°C
Substrate Quantity 1mg

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.