missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Casein, pure

Product Code. 10545691 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic Drum
Plastic bottle
Unit Size:
100g
1kg
25kg
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10545691 100 g Plastic bottle 100g
10063370 1 kg Plastic bottle 1kg
11986941 5 kg Plastic Drum 5kg
15189253 25 kg Fibre drum 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10545691 Supplier Thermo Scientific Chemicals Supplier No. 276071000
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Casein, CAS # 9000-71-9, is a family of related phosphoproteins, usually isolated from bovine milk. | CAS: 9000-71-9 | C81H125N22O39P | 2061.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Information
• Casein contains a mixture of related phosphoproteins present in bovine milk and cheese. It contains all of the common amino acids and is rich in the essential ones.

Applications

• Casein is a source of amino acids, carbohydrates, and two inorganic elements namely calcium and phosphorus
• This compound can be used in vivo as a solid food sample and in vitro digestion experiments with rodents
• It can increase the photostability of quantum dots and decrease nonspecific binding in immunofluorescence experiments
• Casein, in biochemical experiments, can be used to prepare P407-casein hydrogels and to study the mechanical effects of the addition of casein to P407

Literature References

Tundisi, L.; Yang, R.; Borelli, L.; Alves, T.; Mehta, M.; Chaud, M.; Mazzola, P.; Kohane, D. Enhancement of the mechanical and drug-releasing properties of poloxamer 407 hydrogels with casein. Pharm Res. 2021, 38 (3), 515–522.

Irshad, I.; Kanekanian, A.; Peters, A.; Masud, T. Antioxidant activity of bioactive peptides derived from bovine casein hydrolysate fractions. J. Food Sci. Technol. 2015, 52 (1), 231-239.

Sharma, A. K.; Sawhney, I. K. Modelling moisture sorption characteristics in dried acid casein using connectionist paradigm vis-à-vis classical methods. J. Food Sci. Technol. 2015, 52 (1), 151-160.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9000-71-9
Molecular Formula C81H125N22O39P
Molecular Weight (g/mol) 2061.98
MDL Number MFCD00081481
InChI Key BECPQYXYKAMYBN-UHFFFAOYNA-N
Synonym casein, caseinogen, casein, tech., bovine beta-casein monophosphopeptide
PubChem CID 73995022
IUPAC Name 6-amino-2-{[2-({2-[(2-{[2-({2-[(2-{[2-({2-[(2-{[2-({2-[(2-{[2-({2-[(2-amino-1-hydroxy-3-phenylpropylidene)amino]-1-hydroxy-4-(C-hydroxycarbonimidoyl)butylidene}amino)-1-hydroxy-3-(phosphonooxy)propylidene]amino}-4-carboxy-1-hydroxybutylidene)amino]-4-carboxy-1-hydroxybutylidene}amino)-1-hydroxy-4-(C-hydroxycarbonimidoyl)butylidene]amino}-1-hydroxy-4-(C-hydroxycarbonimidoyl)butylidene)amino]-1-hydroxy-4-(C-hydroxycarbonimidoyl)butylidene}amino)-1,3-dihydroxybutylidene]amino}-4-carboxy-1-hydroxybutylidene)amino]-3-carboxy-1-hydroxypropylidene}amino)-4-carboxy-1-hydroxybutylidene]amino}-1-hydroxy-4-methylpentylidene)amino]-1-hydroxy-4-(C-hydroxycarbonimidoyl)butylidene}amino)-3-carboxy-1-hydroxypropylidene]amino}hexanoic acid
SMILES CC(C)CC(N=C(O)C(CCC(O)=O)N=C(O)C(CC(O)=O)N=C(O)C(CCC(O)=O)N=C(O)C(N=C(O)C(CCC(O)=N)N=C(O)C(CCC(O)=N)N=C(O)C(CCC(O)=N)N=C(O)C(CCC(O)=O)N=C(O)C(CCC(O)=O)N=C(O)C(COP(O)(O)=O)N=C(O)C(CCC(O)=N)N=C(O)C(N)CC1=CC=CC=C1)C(C)O)C(O)=NC(CCC(O)=N)C(O)=NC(CC(O)=O)C(O)=NC(CCCCN)C(O)=O

Specifications

Packaging Plastic bottle
Quantity 100 g
Grade Pure
Chemical Name or Material Casein, tech.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.