missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CASK, GST Tag Recombinant Protein

Product Code. 15488923
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
This item is not returnable. View return policy

Product Code. 15488923

Brand: Invitrogen™ PV6271

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Recombinant human CASK (1-570) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. Validated in LanthaScreen™ Kinase Binding Assay Protocols available for simple use and optimized performance Multiple pack sizes available for convenient use Related Product:LanthaScreen™ Eu Kinase Binding Assay Protein Form: Recombinant, Fragment Assay Entry: Biochemical binding/kinetic.

CASK is a calcium/calmodulin-dependent serine protein kinase which is a MAGUK (membrane-associated guanylate kinase) protein family member. CASK is a scaffold protein and the encoded protein is located at synapses in the brain. CASK associates with FG syndrome 4, mental retardation, microcephaly with pontine and cerebellar hypoplasia, and a form of X-linked mental retardation. CASK functions as a cytoskeletal membrane scaffold that coordinates signal transduction pathways within the cortical cytoskeleton.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O14936
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris HCl with 25% glycerol, 0.1mM PMSF, 0.1mM EDTA, 10mM glutathione, 150mM NaCl, 0.25mM DTT and no preservative; pH 7.5
Gene ID (Entrez) 8573
Molecular Weight (g/mol) 91 kDa
Name Human CASK, GST Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias CAGH39; calcium/calmodulin dependent serine protein kinase; calcium/calmodulin-dependent serin protein kinase; Calcium/calmodulin-dependent serine protein kinase; calcium/calmodulin-dependent serine protein kinase (MAGUK family); calcium/calmodulin-dependent serine protein kinase membrane-associated guanylate kinase; CAMGUK; CASK; CMG; DXPri1; DXRib1; FGS4; G-protein-coupled receptor GPR34; hCASK; LIN2; LIN-2; MGC7449; MICPCH; mLin-2; MRXSNA; OTTHUMP00000023157; OTTHUMP00000023161; OTTHUMP00000023162; Pals3; peripheral plasma membrane protein CASK; Protein lin-2 homolog; RP23-278L4.1; TNRC8; trinucleotide repeat containing 8
Common Name CASK
Gene Symbol Cask
Protein Length 1-570
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence 1-570
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Assay Entry Biochemical binding/kinetic
Kinase Group Serine/Threonine Kinase
Target Entry CASK
Expression System Baculovirus
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Fragment
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.