missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Caspase-3 Inhibitor III

Product Code. 15456389
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15456389 1 mg 1mg
15446389 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15456389 Supplier Thermo Scientific Chemicals Supplier No. J64442.M^
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

A potent, cell-permeable, and irreversible inhibitor of caspase-3; also inhibits caspases -6, -7, -8, and -10. Commonly used at concentrations up to 100 -M to examine the role of caspase-3-dependent apoptosis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 285570-60-7
Molecular Formula C21H31ClN4O11
Synonym Ac-DEVD-CMK; N-Acetyl-Asp-Glu-Val-Asp chloromethyl ketone

Specifications

Quantity 1 mg
Description Off-white to brown
Content And Storage -30°C to -10°C
For Use With (Application) A potent, cell-permeable, and irreversible inhibitor of caspase-3; also inhibits caspases -6, -7, -8, and -10. Commonly used at concentrations up to 100 →M to examine the role of caspase-3-dependent apoptosis
Formula Weight 550.94
Solubility Soluble in DMSO
Form Powder
InChI Key ATNOUPFYBMVFLD-UHFFFAOYNA-N
SMILES CC(C)C(NC(=O)C(CCC(O)=O)NC(=O)C(CC(O)=O)NC(C)=O)C(=O)NC(CC(O)=O)C(=O)CCl
IUPAC Name 3-{2-[4-carboxy-2-(3-carboxy-2-acetamidopropanamido)butanamido]-3-methylbutanamido}-5-chloro-4-oxopentanoic acid
Molecular Weight (g/mol) 550.95
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.