missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Caspase-3 Substrate III, fluorogenic

Product Code. 15468419
Change view
Click to view available options
Substrate Quantity:
0.5mg
1mg
Unit Size:
0.5mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Substrate Quantity Unit Size
15468419 0.5mg 0.5mg
15478419 1mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15468419 Supplier Thermo Scientific Chemicals Supplier No. J66881.!D
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For cellular and molecular biology applications

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Color White to off-white
Form Lyophilized
Substrate Type Caspase Substrate
Format null
Molecular Weight (g/mol) 1155.08
Storage Requirements -30° to -10°C
Substrate Quantity 0.5mg

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.