missing translation for 'onlineSavingsMsg'
Learn More

Caspase-9 Inhibitor III, Thermo Scientific Chemicals

Product Code. 15456939
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15456939 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15456939 Supplier Thermo Scientific Alfa Aesar Supplier No. J65501.M^

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A potent and irreversible inhibitor of caspase-9

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 403848-57-7
Molecular Formula C24H35ClN6O9
Synonym Ac-LEHD-CMK;

Specifications

Quantity 1 mg
Description White
Content And Storage -30°C to -10°C
For Use With (Application) A potent and irreversible inhibitor of caspase-9
Formula Weight 587.02
Form Powder

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.