missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cassette Clamp, left

Product Code. 15324574
Change view
Click to view available options
Quantity:
1 unit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15324574 1 unit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15324574 Supplier Invitrogen™ Supplier No. A25946
  • ONLINE EXCLUSIVE PRICE
    146.00 €
    118.26 € / 1 units
    Save 27.74 €
    19% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cassette clamps are replacement parts for the Mini Gel Tank. They lock and seal the gel cassettes within the Gel Runner Tank. The orientation is determined when facing the front of the gel tank.

About the Mini Gel Tank
The Mini Gel Tank is compatible with a variety of Novex™ gels, NuPAGE™ gels, and Bolt™ Bis-Tris Plus gels. Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient side-by-side gel loading and enhanced viewing during use. Optimized conditions using constant voltage allow for Bolt™ Bis-Tris Plus gels to be run in approximately 35 minutes. Run times may vary depending on buffer conditions and power supplies being used.

For Research Use Only. Not for use in diagnostic procedures.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Universal
Gel Compatibility Novex™ Mini Gels
Gel Size Mini
Type Cassette Clamp
Quantity 1 unit
Shipping Condition Room Temperature
Can I purchase replacement parts for the Mini Gel Tank?

Here are the replacement parts we offer for the Mini Gel Tank:
Replacement part - Cat. No.
Gel Runner Tank - B4478641
Mini Gel Tank Lid - A25944
Mini Gel Tank Base - A25950
Cassette Clamp, left - A25946
Cassette Clamp, right - A25945
Cassette Clamp Cam Handle Set - A26732

What are the components of the Mini Gel Tank?

The Mini Gel Tank includes the Mini Gel Electrophoresis Tank, Mini Gel Tank Lid, Cassette Clamp (Right), 10 cm, Cassette Clamp (Left), 10 cm, Mini Gel Tank Base, Invitrogen Power Supply Adapters, and Gel Knife.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.