missing translation for 'onlineSavingsMsg'
Learn More

LP Italiana Spa™ Embedding Cassette with Lid

Product Code. 11886085 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
Embedding Cassette
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
11886085 Embedding Cassette Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11886085 Supplier LP Italiana Spa™ Supplier No. 191022
  • ONLINE EXCLUSIVE PRICE
    689.00 €
    616.00 € / Pack of 1
    Save 73.00 €
    10% Off
 Restricted Item
Estimated Shipment: 01-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

White embedding cassette with lid for pathological embedding.

White embedding cassette with lid for pathological embedding.

Embedding cassette for use in the pathological embedding proccess.

  • With lid
  • No ring
  • Great for use with tissue samples
TRUSTED_SUSTAINABILITY

Specifications

Product Type Embedding Cassette
Color White
For Use With (Application) Embedding
Includes Lid
Quantity 250 Pk.
Weight (English) 0.13 oz.
Weight (Metric) 3.72 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.