missing translation for 'onlineSavingsMsg'
Learn More

Castanospermine, MedChemExpress

Product Code. 16489248
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16489248 10 mg 10mg
16474927 50 mg 50mg
16429176 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16489248 Supplier Medchem Express Supplier No. HYN2022
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Castanospermine inhibits all forms of α- and β-glucosidases, especially glucosidase l (required for glucoprotein processing by transfer of mannose and glucose from asparagine-linked lipids).

Castanospermine inhibits all forms of α- and β-glucosidases, especially glucosidase l (required for glucoprotein processing by transfer of mannose and glucose from asparagine-linked lipids).

Specifications

Chemical Name or Material Castanospermine
CAS 79831-76-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C8H15NO4
Solubility Information H2O : 100 mg/mL (528.51 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (528.51 mM; Need ultrasonic)
SMILES O[C@@H]1[C@@H](O)[C@H](O)[C@]([C@@H](O)CC2)([H])N2C1
Molecular Weight (g/mol) 189.21
Quantity 10 mg
Formula Weight 189.21
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.