missing translation for 'onlineSavingsMsg'
Learn More

Castanospermine, Tocris Bioscience™

Product Code. 18015072
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18015072 10 mg 10mg
18035672 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18015072 Supplier Tocris Bioscience Supplier No. 0759/10

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Glucosidases α and β inhibitor

Potent inhibitor of α- and β-glucosidases, especially glucosidase l (required for glucoprotein processing by transfer of mannose and glucose from asparagine-linked lipids). Inhibits HIV syncytium formation and replication.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79831-76-8
Molecular Formula C8H15NO4
Molecular Weight (g/mol) 189.211
InChI Key JDVVGAQPNNXQDW-TVNFTVLESA-N
Synonym castanospermine, castinospermine, 1,6,7,8-tetrahydroxyoctahydroindolizine, 1s,6s,7r,8r,8ar-1,6,7,8-tetrahydroxyindolizidine, 6-epicastanospermine, unii-q0i3184xm7, 6,7-diepicastanospermine, castanospermine, castanospermum australe, 1s,6s,7r,8ar-tetrahydroxyoctahydroindolizine, 1s,6s,7r,8r,8ar-octahydroindolizine-1,6,7,8-tetrol
PubChem CID 54445
ChEBI CHEBI:27860
IUPAC Name (1S,6S,7R,8R,8aR)-1,2,3,5,6,7,8,8a-octahydroindolizine-1,6,7,8-tetrol
SMILES C1CN2CC(C(C(C2C1O)O)O)O

Specifications

Quantity 10 mg
Formula Weight 189.21
Percent Purity >98%
Chemical Name or Material Castanospermine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.