missing translation for 'onlineSavingsMsg'
Learn More

Castor oil

Product Code. 10539813 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
500 g
1 kg
2.5 kg
Unit Size:
1kg
2.5kg
250g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10539813 2.5 kg 2.5kg
10295234 250 g 250g
10197170 500 g 500g
10422514 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10539813 Supplier Thermo Scientific Acros Supplier No. 404160025
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 8001-79-4 | C57H104O9 | 933.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00130879

Specifications

Melting Point -10°C
Color Yellow
Density 0.9570g/mL
Boiling Point 229°C
Flash Point 229°C
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4770 to 1.481
Quantity 2.5 kg
Saponification Value 176 to 182g KOH/g
Merck Index 14,1898
Specific Gravity 0.957
Specific Rotation + 3.5
Solubility Information Solubility in water: .. Other solubilities: miscible with alcohol,benzene,chloroform,,carbondisulfide,ether and glacial acetic acid
Viscosity 0,6-0,8 Pas (25°C)
Physical Form Viscous Liquid
Chemical Name or Material Castor oil
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.