missing translation for 'onlineSavingsMsg'
Learn More

Catecholborane, 97%

Product Code. 11318106
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11318106 25 g 25g
11308106 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11318106 Supplier Thermo Scientific Alfa Aesar Supplier No. L14998.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 274-07-7 | C6H4BO2 | 118.906 g/mol

Catecholborane is used to prepare B-alkylcatecholboranes. It finds application for the preparation of amides and macrocyclic lactams from carboxylic acids. It is used as a stereoselective reducing agent to convert beta-hydroxy ketones to syn 1,3-diols. Further, it reacts with alkyne through hydroboration to form trans vinyl borane, which is a precursor to Suzuki reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Catecholborane is used to prepare B-alkylcatecholboranes. It finds application for the preparation of amides and macrocyclic lactams from carboxylic acids. It is used as a stereoselective reducing agent to convert beta-hydroxy ketones to syn 1,3-diols. Further, it reacts with alkyne through hydroboration to form trans vinyl borane, which is a precursor to Suzuki reaction.

Solubility
Miscible with diethyl ether, tetrahydrofuran, dichloromethane, chloroform, carbon tetrachloride, toluene, and benzene.

Notes
Moisture sensitive. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 274-07-7
Molecular Formula C6H4BO2
Molecular Weight (g/mol) 118.906
MDL Number MFCD00005846
InChI Key ZDQWVKDDJDIVAL-UHFFFAOYSA-N
Synonym catecholborane, 1,3,2-benzodioxaborole, benzo d 1,3,2 dioxaborole, 1,3,2$l^ 2-benzodioxaborole, cenmejuyoomffz-uhfffaoysa-n, catechol borane, catecholatoborane, benzcatechinborane, pyrocatecholborane
PubChem CID 6327445
IUPAC Name 1,3,2$l^{2}-benzodioxaborole
SMILES [B]1OC2=CC=CC=C2O1

Specifications

Melting Point 12°C
Density 1.125
Boiling Point 50°C (50 mmHg)
Flash Point 2°C (35°F)
Quantity 25 g
UN Number UN2924
Beilstein 972072
Sensitivity Moisture sensitive; May form a precipitate
Solubility Information Miscible with diethyl ether,tetrahydrofuran,dichloromethane,chloroform,carbon tetrachloride,toluene,and benzene.
Formula Weight 119.92
Percent Purity 97%
Chemical Name or Material Catecholborane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.