missing translation for 'onlineSavingsMsg'
Learn More

Catecholborane (Technical Grade), TRC

Product Code. 30368594
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30368594 5 g 5g
30365388 1 g 1g
30367656 10 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30368594 Supplier Toronto Research Chemicals Supplier No. TRCC2151855G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 274-07-7
Molecular Formula C6H5BO2
Molecular Weight (g/mol) 119.91
Synonym 1,3,2-Benzodioxaborole, Benzcatechinborane, CB, Catecholatoborane, Pyrocatecholborane
IUPAC Name 1,3,2-benzodioxaborole
SMILES B1Oc2ccccc2O1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C6H5BO2/c1-2-4-6-5(3-1)8-7-9-6/h1-4,7H
Formula Weight 120.0383
Chemical Name or Material Catecholborane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.