missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cathepsin G Inhibitor I

Product Code. 15427149
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Product Code. Quantity unitSize
15427149 1 mg 1mg
1 options
This item is not returnable. View return policy

Product Code. 15427149

Brand: Thermo Scientific Chemicals J65957.M^

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

A potent, selective, reversible, competitive, non-peptide inhibitor of cathepsin G

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 mg
Description White
Content And Storage -30°C to -10°C
For Use With (Application) A potent, selective, reversible, competitive, non-peptide inhibitor of cathepsin G
CAS 429676-93-7
Molecular Formula C36H33N2O6P
Formula Weight 620.60
Sensitivity Light Sensitive
Solubility Soluble in DMSO
Form Solid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.