missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cathepsin K Substrate (fluorogenic)

Product Code. 15456189
Change view
Click to view available options
Substrate Quantity:
10mg
25mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Substrate Quantity unitSize
15456189 25mg 25mg
15446189 10mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15456189 Supplier Thermo Scientific Chemicals Supplier No. J64082.MB
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

For cellular and molecular biology applications | 578.65 g/mol

A sensitive fluorogenic substrate for cathepsin K

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Color White
Form Solid
For Use With (Application) A sensitive fluorogenic substrate for cathepsin K
Substrate Type Cathepsin Substrate
Molecular Weight (g/mol) 578.65
Storage Requirements -30° to -10°C
Substrate Quantity 25mg

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.