missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CB 13

Product Code. 18063892
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18063892

Brand: Tocris Bioscience™ 2928/50

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Potent dual CB1/CB2 agonist

CB 13 is a potent, orally active CB1/CB2 receptor agonist with limited brain penetration (EC50 values are 6.1 and 27.9 nM for CB1 and CB2 receptors respectively). Displays antihyperalgesic activity in a rat model of neuropathic pain with no CNS side effects.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Naphthalenyl[4-(pentyloxy)-1-naphthalenyl]methanone
CAS 432047-72-8
Quantity 50 mg
Target Non-selective Cannabinoid Receptor Agonists
Molecular Formula C26H24O2
Purity 0.99
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.