missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CB 13
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
CB 13 is a potent, orally active CB1/CB2 receptor agonist with limited brain penetration (EC50 values are 6.1 and 27.9 nM for CB1 and CB2 receptors respectively). Displays antihyperalgesic activity in a rat model of neuropathic pain with no CNS side effects.
Specifications
Specifications
| Chemical Name or Material | 1-Naphthalenyl[4-(pentyloxy)-1-naphthalenyl]methanone |
| CAS | 432047-72-8 |
| Quantity | 50 mg |
| Target | Non-selective Cannabinoid Receptor Agonists |
| Molecular Formula | C26H24O2 |
| Purity | 0.99 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction