missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CB-5083, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
CB-5083 is a first-in-class, potent, selective, and orally bioavailable inhibitor of the p97 AAA ATPase/VCP. CB-5083 selectively inhibits p97 through its D2 site with the IC50 of 11 nM。
Specifications
Specifications
| Chemical Name or Material | CB-5083 |
| CAS | 1542705-92-9 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C24H23N5O2 |
| Quantity | 100 mg |
| Solubility Information | DMSO : 100 mg/mL (241.86 mM; Need ultrasonic) |
| SMILES | O=C(C1=CC=CC2=C1C=C(C)N2C3=NC(NCC4=CC=CC=C4)=C(COCC5)C5=N3)N |
| Molecular Weight (g/mol) | 413.47 |
| Formula Weight | 413.47 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction