missing translation for 'onlineSavingsMsg'
Learn More

CBZ-L-Serine methyl ester, 95%, Thermo Scientific Chemicals

Product Code. p-4228491 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10321800 1 g 1g
10230000 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10321800 Supplier Thermo Scientific Acros Supplier No. 434950010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1676-81-9
Molecular Formula C12H15NO5
Molecular Weight (g/mol) 253.25
MDL Number MFCD00077039
InChI Key CINAUOAOVQPWIB-UHFFFAOYNA-N
Synonym z-ser-ome, n-cbz-l-serine methyl ester, cbz-ser-ome, n-carbobenzoxy-l-serine methyl ester, z-l-serine methyl ester, cbz-l-serine methyl ester, n-z-l-serine methyl ester, s-methyl 2-benzyloxy carbonyl amino-3-hydroxypropanoate, n-carbobenzyloxy-l-serine methyl ester, d-n-cbz-serine methyl ester
PubChem CID 6999535
IUPAC Name methyl (2S)-3-hydroxy-2-(phenylmethoxycarbonylamino)propanoate
SMILES COC(=O)C(CO)NC(=O)OCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 170°C (0.1 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Specific Rotation -13.5
Formula Weight 253.26
Percent Purity 95%
Physical Form Viscous Liquid or Solid
Chemical Name or Material CBZ-L-Serine methyl ester
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.