missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CC4

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
CC4 is a high affinity and subtype selective α6β2 and α4β2 partial agonist (Ki values are 12 and 26nM for rat α6β2 and α4β2 receptors respectively). Has low affinity for α3β4 and α7 receptors (Ki values are 4.8 and 13 μM for human α3β4 and rat α7 receptors respectively). Stimulates dopamine release from striatal slices in vitro. Attenuates nicotine-induced self-administration and conditional place preference in rats.
Specifications
Specifications
| Chemical Name or Material | (1R,5S)-1,2,3,4,5,6-Hexahydro-3-[2-[(1R,5S)-1,5,6,8-tetrahydro-8-oxo-1,5-methano-2H-pyrido[1,2-a][1,5]diazocin-3(4H)-yl]ethyl]-1,5-methano-8H-pyrido[1,2-a][1,5]diazocin-8-one |
| CAS | 492-02-4 |
| Quantity | 10 mg |
| Target | Additional Nicotinic (a4b2) Receptor Compounds |
| Molecular Formula | C24H30N4O2 |
| Formula Weight | 406.52 |
| Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction