missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ CCCP

Product Code. 18782524
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18782524 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18782524 Supplier R&D Systems™ Supplier No. 0452/500
  • ONLINE EXCLUSIVE PRICE
    217.92 €
    194.25 € / 500mg
    Save 23.67 €
    10% Off
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Oxidative phosphorylation uncoupler

Widely used uncoupler of oxidative phosphorylation.
TRUSTED_SUSTAINABILITY

Specifications

CAS 555-60-2
Quantity 500 mg
Product Line Tocris
Molecular Formula C9H5ClN4
Formula Weight 204.62
Purity >98%
Molecular Weight (g/mol) 204.617
Chemical Name or Material CCCP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.