missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCF2-FA (free acid)
Description
CCF2-FA (Free Acid) is a fluorescence resonance energy transfer (FRET) substrate that consists of a cephalosporin core linking B7-hydroxycoumarin to fluorescein. This product is intended for use as a beta-lactamase substrate in a GeneBLAzer™ cell-based assay.
GeneBLAzer™ cell-based assays typically utilize the membrane-permeant ester forms (CCF2-AM and CCF4-AM) of the negatively charged fluorescent beta-lactamase substrates, CCF2 and CCF4. These lipophilic esters readily enter the cell, where cleavage by endogeneous cytoplasmic esterases rapidly converts them into their negatively charged forms, thereby trapping them in the cytosol.
CCF2-FA is essentially the CCF2 substrate without the esters found in the AM version. CCF2-FA is de-esterified and used in cell lysate applications, bypassing loading across the cell membrane and de-esterification steps. Cell lysates are the preferred method for applications using cells that contain a cell wall. CCF2-FA can also be used as a control to acquire the excitation and emission spectra for CCF2-AM and CCF4-AM.
GeneBLAzer™ cell-based assays typically utilize the membrane-permeant ester forms (CCF2-AM and CCF4-AM) of the negatively charged fluorescent beta-lactamase substrates, CCF2 and CCF4. These lipophilic esters readily enter the cell, where cleavage by endogeneous cytoplasmic esterases rapidly converts them into their negatively charged forms, thereby trapping them in the cytosol.
CCF2-FA is essentially the CCF2 substrate without the esters found in the AM version. CCF2-FA is de-esterified and used in cell lysate applications, bypassing loading across the cell membrane and de-esterification steps. Cell lysates are the preferred method for applications using cells that contain a cell wall. CCF2-FA can also be used as a control to acquire the excitation and emission spectra for CCF2-AM and CCF4-AM.
Specifications
Specifications
| Cell Permeability | Cell-Impermeant |
| Content And Storage | Store in freezer (-5 to -30°C). |
| Detection Method | Fluorescence |
| Product Line | GeneBLAzer |
| Quantity | 1mg |
| Substrate | CCF2-FA |
| Substrate Properties | Chemical Substrate |
| Substrate Type | Beta-Lactamase Substrate |
| Shipping Condition | Approved for shipment on Wet or Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?