missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL28 Rat anti-Mouse, Clone: 134315, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB5331-SP
This item is not returnable.
View return policy
Description
CCL28 Monoclonal specifically detects CCL28 in Mouse samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
| CCL28 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CCL28, chemokine (C-C motif) ligand 28, MEC, member 28, MGC71902, Protein CCK1, VIC | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 56477 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| ELISA | |
| 134315 | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Q9JIL2 | |
| CCL28 | |
| E. coli-derived recombinant mouse CCL28 Ile23-Arg130 Accession # Q9JIL2 | |
| 25 μg | |
| Primary | |
| Detects mouse CCL28 in ELISAs. In sandwich immunoassays, no cross-reactivity with recombinant human CCL28 or recombinant mouse CCL27 is observed. | |
| Mouse | |
| Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?