missing translation for 'onlineSavingsMsg'
Learn More

CCT 137690, MedChemExpress

Product Code. 16466877
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16466877 5 mg 5mg
16425136 100 mg 100mg
16404178 10 mg 10mg
16400527 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16466877 Supplier Medchem Express Supplier No. HY108045mg
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress CCT 137690 is a potent and orally available aurora kinase inhibitor with IC50s of 15, 25, and 19 nM for aurora A, B and C, respectively.

CCT 137690 is a potent and orally available aurora kinase inhibitor with IC50s of 15, 25, and 19 nM for aurora A, B and C, respectively.

Specifications

Chemical Name or Material CCT 137690
CAS 1095382-05-0
Color White
Purity Grade Notes Research
Molecular Formula C26H31BrN8O
Quantity 5 mg
Solubility Information DMSO : 16.67 mg/mL (30.23 mM; Need ultrasonic)
SMILES CC1=CC(CN(CC2)CCN2C3=C4C(NC(C5=CC=C(N6CCN(C)CC6)C=C5)=N4)=NC=C3Br)=NO1
Molecular Weight (g/mol) 551.48
Formula Weight 551.48
Percent Purity 99.1%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.