missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CCT 241533 dihydrochloride

Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
CCT 241533 dihydrochloride is a potent Chk2 inhibitor (IC50 = 3 nM). Shows >63-fold selectivity for Chk1 over Chk2 and a panel of 84 other kinases. Inhibits Chk2 activation in response to etoposide -induced DNA damage in HT29 cells. Blocks ionizing radiation-induced apoptosis of mouse thymocytes.
Specifications
Specifications
| Chemical Name or Material | (3R,4S)-4-[[2-(5-Fluoro-2-hydroxyphenyl)-6,7-dimethoxy-4-quinazolinyl]amino]-α,α-dimethyl-3-pyrrolidinemethanol dihydrochloride |
| CAS | 1962925-28-5 |
| Quantity | 10 mg |
| Target | Checkpoint Kinase Inhibitors |
| Molecular Formula | C23H27FN4O4.2HCl |
| Formula Weight | 515.41 |
| Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction