missing translation for 'onlineSavingsMsg'
Learn More

Ytterbium(III) 2,4-pentanedionate, REacton™, 99.9% (REO)

Product Code. 11392258
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11392258 10 g 10g
11302268 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11392258 Supplier Thermo Scientific Alfa Aesar Supplier No. 013159.09
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14284-98-1 | C15H21O6Yb | 470.37 g/mol

Ytterbium(III) 2,4-pentanedionate is used as a catalyst for the preparation of aliphatic oligocarbonate polyols. It takes part in transesterification reactions. It is used for the preparation of polyesters or polyacrylates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14284-98-1
Molecular Formula C15H21O6Yb
Molecular Weight (g/mol) 470.37
MDL Number MFCD00013508
InChI Key JKJCKIFLFXFUKJ-UHFFFAOYSA-N
Synonym ytterbium-2,4-pentanedionate, ytterbium acetyl acetonate
PubChem CID 14455610
IUPAC Name (Z)-4-oxopent-2-en-2-olate;ytterbium(3+)
SMILES [Yb+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Specifications

Color White
Odor Odorless
Quantity 10 g
Solubility Information Insoluble in water.
Formula Weight 470.37
Percent Purity 99.9%
Grade Reacton
Physical Form Powder
Assay (REO)
Chemical Name or Material Ytterbium(III) 2,4-pentanedionate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.