missing translation for 'onlineSavingsMsg'
Learn More

CD CHO Medium

Product Code. 16453869
Change view
Click to view available options
Quantity:
10 L
Unit Size:
10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16453869 10 L 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16453869 Supplier Invitrogen Supplier No. 10743001
 Restricted Item
Estimated Shipment: 21-08-2026
Añadir a la cesta
Añadir a la cesta
Este artículo no se puede devolver. Vea la política de devoluciones

Especificaciones

Cell Line CHO
Cell Type Ovarian Cells
Classification Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Concentration 1 X
Form Liquid
Product Type CD CHO Medium
Serum Level Serum-free
Sterility Sterile-filtered
With Additives Sodium Pyruvate, Sodium Bicarbonate, Non Essential Amino Acids (NEAA)
Without Additives No Glutamine, No Phenol Red, No Thymidine, No Hypoxanthine
Product Line Gibco
Quantity 10 L
Shipping Condition Room Temperature
Endotoxin Level Low
Format Universal Bag
Included Antibiotics No antibiotics
Purity or Quality Grade Cell Culture Grade
Concentration Ratio 1 :1
Content And Storage Store at 2-8°C in the dark.
Culture Environment CO2
Culture Type Suspension Cell Culture
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.