missing translation for 'onlineSavingsMsg'
Learn More

CD CHO Medium

Product Code. 17285946 Shop All Gibco Products
Change view
Click to view available options
Quantity:
10 L
20 L
Unit Size:
10 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17285946 20 L 20 L
16453869 10 L 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17285946 Supplier Gibco Supplier No. 10743002
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Cell Line CHO
Cell Type Ovarian Cells
Classification Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Concentration 1 X
Form Liquid
Product Type CD CHO Medium
Serum Level Serum-free
Sterility Sterile-filtered
With Additives Sodium Pyruvate, Sodium Bicarbonate, Non Essential Amino Acids (NEAA)
Without Additives No Glutamine, No Phenol Red, No Thymidine, No Hypoxanthine
Product Line Gibco
Quantity 20 L
Shipping Condition Room Temperature
Endotoxin Level Low
Format Universal Bag
Included Antibiotics No antibiotics
Purity or Quality Grade Cell Culture Grade
Concentration Ratio 1 :1
Content And Storage Store at 2-8°C in the dark.
Culture Environment CO2
Culture Type Suspension Cell Culture
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.