missing translation for 'onlineSavingsMsg'
Learn More

CD11c Mouse, Clone: ITGAX/1242, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio 3687-MSM2-P0

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15920288

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

CD11c
Cell surface
ITGAX/1242
Unconjugated
XP_011544154
Recombinant human ITGAX protein fragment (aa 637-827)
Human dendritic cells. Human lymph nodes and tonsils.
20 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Immunohistochemistry
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% Azide
Mouse
145kDa
Protein A/G Purified
RUO
3687
IgG2b κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers