missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD155/PVR Mouse anti-Human, Alexa Fluor 488, Clone: 300907, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB25301G-100UG
This item is not returnable.
View return policy
Description
CD155/PVR Monoclonal specifically detects CD155/PVR in Human samples. It is validated for Flow Cytometry.Specifications
| CD155/PVR | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| AAH15542 | |
| PVR | |
| Mouse myeloma cell line NS0-derived recombinant human CD155/PVR Gly27-Asn343 Accession # AAH15542 | |
| RUO | |
| 5817 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 300907 | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CD155, CD155 antigen, HVED, Necl-5, NECL5, nectin-like 5, Nectin-like protein 5, poliovirus receptor, PVR, PVS, PVSFLJ25946, Tage4 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human CD155/PVR in direct ELISAs and Western blots. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction