missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BB700 Mouse Anti-Human CD25, Clone M-A251 (RUO)
Description
- The M-A251 monoclonal antibody specifically binds to the 55 kDa type I transmembrane glycoprotein known as low-affinity interleukin-2 receptor alpha chain subunit (IL-2Rα).
- CD25 is expressed on regulatory T cells, activated lymphocytes (T and B), and monocytes.
- It associates with the IL-2Rβ/CD122 and IL-2Rγ/CD132 receptor chains to form the high-affinity IL-2R complex.
- CD25 expression on T and B lymphocytes is upregulated by antigenic or mitogenic stimulation.
- Soluble CD25/IL-2Rα is produced as a consequence of lymphocyte stimulation and is found in biological fluids following inflammatory responses.
- The antibody was conjugated to BD Horizon BB700, which is part of the BD Horizon Brilliant™ Blue family of dyes.
- It is a polymer-based tandem dye developed exclusively by BD Biosciences.
- With an excitation max of 485 nm and an emission max of 693 nm, BD Horizon BB700 can be excited by the 488 nm laser and detected in a standard PerCP-Cy™5.5 set (eg., 695/40-nm filter).
- This dye provides a much brighter alternative to PerCP-Cy5.5 with less cross laser excitation off the 405 nm and 355 nm lasers.
Specifications
Specifications
| Antigen | CD25 (IL-2 Receptor α) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | M-A251 |
| Conjugate | Brilliant Blue 700 (BB700) |
| Host Species | Mouse |
| Quantity | 100 Tests |
| Regulatory Status | RUO |
| Primary or Secondary | Secondary |
| Test Specificity | CD25 (IL-2 Receptor α) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?