missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CD273(PDCD1LG2)/CEN9q FISH Probe
Specifications
Specifications
| Content And Storage | Store at 4°C in the dark |
| Format | Tube |
| For Use With (Application) | Fluorescent In Situ Hybridization (Cell) |
| Gene | programmed cell death 1 ligand 2 |
| Gene Alias | B7DC,Btdc,CD273,MGC142238,MGC142240,PD-L2,PDCD1L2,PDL2,bA574F11.2 |
| Gene Symbol | PDCD1LG2 |
| Gene ID (Entrez) | 80380 |
| Product Type | DNA/RNA |
| Quantity | 200 μL |
| Form | Liquid |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?