missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ CD274(PD-L1)-PDCD1LG2(CD273)/CEN9q FISH Probe

Product Code. 16093365
Change view
Click to view available options
Quantity:
100 μL
200 μL
Unit Size:
100µL
200µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16093365 100 μL 100µL
16003375 200 μL 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16093365 Supplier Abnova™ Supplier No. FG0226.100uL
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.

Specifications

Content And Storage Store at 4°C in the dark
Format Tube
For Use With (Application) Fluorescent In Situ Hybridization (Cell)
Gene CD274 molecule
Gene Alias B7-H,B7H1,MGC142294,MGC142296,PD-L1,PDCD1L1,PDCD1LG1,PDL1
Gene Symbol CD274
Gene ID (Entrez) 29126;80380
Product Type DNA/RNA
Quantity 100 μL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.