missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences CD3/CD62L/CD45RA Fab-TACS™ Magnetic Microbead Starter Kit
Shop All IBA Lifesciences Products
Click to view available options
:
Human
Beschreibung
- Fab-TACS™ - Traceless Affinity Cell Selection - is a positive cell isolation approach based on our Strep-tag™ technology
- CD3/CD62L/CD45RA Fab-TACS™ Magnetic Microbead Starter Kit allows label-free isolation of CD3+ central memory (CD3+CD62L+CD45RA-) or naive (CD3+CD62L+CD45RA+) T cells from peripheral blood mononuclear cells (PBMCs)
- Alternatively, CD3/CD62L/CD45RO Fab-TACS™ Magnetic Microbead Starter Kit is suitable for the isolation of these cells
- Low affinity, Fab fragments fused to a Twin-Strep-tag™ (Fab-Streps) are incubated with Strep-Tactin™ Magnetic Microbeads to form a complex that stably binds to the cell surface
- After specific labeling, target cells are bound to a magnet and non-target cells efficiently washed away
- In a final step, the addition of biotin causes the release of the isolated cells from magnetic microbeads. The Fab-Streps self-dissociate from the cell surface due to their low affinity
- After removing biotin by washing, the isolated cells are ready for further positive selection or depletion steps or can directly be used for further downstream applications
Spezifikation
Spezifikation
| Includes | Strep-Tactin™ Magnetic Microbeads, CD3/CD62L/CD45RA Fab-Strep (human), 10 x Buffer CI, Biotin stock solution |
| Target Species | Human |
| For Use With (Application) | Label-Free Isolation of Naive or Central Memory T Cells From PBMCs |
For Research Use Only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction