Learn More
IBA Lifesciences CD3 Isolation Kit for FABian™
IBA Lifesciences™ CD3 Isolation Kit for FABian™ is for reversible cell isolation delivering label-free target cells.
Brand:  IBA Lifesciences 6-6001-010
Additional Details : Weight : 1.70000kg
 View more versions of this product
Product Code. 15839774
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • The kit is used for the selection of CD3+ T cells from whole blood or buffy coat in conjunction with the FABian™ system.
  • It includes CD3 Fab-Strep (human), the Cell Isolation Column, Cell Isolation Buffer, Biotin for Cell Elution and tubes and a luer lock adapter with the syringe.
  • Each kit is sufficient for one FABian™ run to isolate cells from 9mL whole blood.
Specifications
Specifications
Isolation of label-free cells
2°C to 8°C
9 mL Whole Blood
Non-magnetic Cell Isolation Kit
Human Whole Blood
10 Kits, Human