Learn More
CD36/SR-B3 Goat anti-Mouse, Polyclonal, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody has been used in 24 publications
Brand:  R&D Systems AF2519-SP
Encompass
Product Code. 15701514
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
CD36/SR-B3 Polyclonal specifically detects CD36/SR-B3 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).