missing translation for 'onlineSavingsMsg'
Learn More

CD5 Mouse, PE, Clone: UCHT2, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio QAB15-PE-25Tests

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15961548

  • 119.00 € / 25 tests
Estimated Shipment: 12-09-2024
to see stock.

Description

Description

Specifications

Specifications

CD5
Monoclonal
5μL/Test
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
CD5
Protein A/G Purified
RUO
921
IgG1 κ
Flow Cytometry
UCHT2
PE
NP_055022
Mouse
25 Tests
Primary
Store at 2-4°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers