missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BB700 Mouse Anti-Human CD56 Clone NCAM16.2 (NCAM16) (RUO)
Description
- The NCAM16.2 monoclonal antibody specifically binds to human CD56.
- It recognizes an extracellular immunoglobulin-like domain common to 120, 140, and 180 kDa forms of CD56, also known as the neural cell adhesion molecule (NCAM), NKH1 or MSK39.
- The CD56 antigen is expressed on approximately 10% to 25% of peripheral blood lymphocytes.
- It is present on essentially all resting and activated CD16+ natural killer (NK) lymphocytes and approximately 5% of CD3+ peripheral blood lymphocytes. CD3+ CD56+ T lymphocytes comprise a unique subset of cytotoxic T lymphocytes that mediates non-major histocompatibility complex (MHC)-restricted cytotoxicity.
- CD56 antigen density on NK lymphocytes increases upon cellular activation.
- The CD56 antigen is involved in neuronal homotypic cell adhesion and cell differentiation during embryogenesis.
- CD16+ CD56+ NK cells demonstrate reciprocal transfer of an activation state with dendritic cells.
- The antibody was conjugated to BD Horizon BB700, which is part of the BD Horizon Brilliant™ Blue family of dyes.
- It is a polymer-based tandem dye developed exclusively by BD Biosciences.
- With an excitation max of 485 nm and an emission max of 693 nm, BD Horizon BB700 can be excited by the 488 nm laser and detected in a standard PerCP-Cy™5.5 set (eg, 695/40-nm filter).
- This dye provides a much brighter alternative to PerCP-Cy5.5 with less cross laser excitation off the 405 nm and 355 nm lasers.
Specifications
Specifications
| Antigen | CD56 (NCAM-1) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | NCAM16.2 (also known as NCAM 16) |
| Conjugate | Brilliant Blue 700 (BB700) |
| Host Species | Mouse |
| Quantity | 100 Tests |
| Regulatory Status | RUO |
| Primary or Secondary | Secondary |
| Test Specificity | CD56 (NCAM-1) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?