missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8a Monoclonal Antibody (OKT8 (OKT-8)), NovaFluor™ Blue 610-30S, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
182.00 € - 402.00 €
Specifications
| Antigen | CD8a |
|---|---|
| Clone | OKT8 (OKT-8) |
| Concentration | 4 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
CD8a Monoclonal antibody specifically detects CD8a in Human samples. It is validated for Flow CytometrySpecifications
| CD8a | |
| 4 μL/Test | |
| Monoclonal | |
| Mouse | |
| Human | |
| P01732, P10966 | |
| 925, 926 | |
| Primary | |
| 4° C, store in dark, DO NOT FREEZE! |
| OKT8 (OKT-8) | |
| Flow Cytometry | |
| NovaFluor Blue 610-30S | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| BB154331, CD8, CD8 alpha, CD8 alpha chain, CD8 alpha chain precursor, CD8 antigen, CD8 antigen 32 kDa chain, CD8 antigen 37 kDa chain, CD8 antigen alpha polypeptide, CD8 antigen alpha protein, CD8 antigen alpha protein precursor, CD8 antigen alpha-chain, CD8 antigen beta polypeptide, CD8 antigen beta polypeptide precursor, CD8 antigen beta-chain, CD8 antigen, alpha chain, CD8 antigen, alpha polypeptide, CD8 antigen, alpha polypeptide (p32), CD8 antigen, alpha-chain, CD8 antigen, beta chain, CD8 antigen, beta chain 1, CD8 antigen, beta polypeptide, CD8 antigen, beta polypeptide 1 (p37), CD8 antigen, beta-chain, CD8 beta, CD8 beta chain, CD8 beta-2, CD8a, CD8A antigen alpha, CD8a molecule, CD8A, T-cell surface glycoprotein, CD8alpha, CD8B, CD8b antigen, CD8b molecule, CD8b molecule pseudogene, Cd8b1, CD8beta, CD8BP, fCD8, LEU2, Leu-2, Leu2 T-lymphocyte antigen, leu-2a, Ly-2, LY3, Ly-3, Ly-35, Ly-B, Ly-C, Lymphocyte antigen 3, Lyt2, Lyt-2, Lyt-2.1 lymphocyte differentiation antigen (AA at 100), Lyt3, Lyt-3, MAL, membrane glycoprotein, membrane protein, OKT8 T-cell antigen, OX-8 membrane antigen, p32, P37, RHACD8-4, T cell co-receptor, T lymphocyte surface glycoprotein beta chain, T8 T-cell antigen, T-cell antigen Leu2, T-cell membrane glycoprotein Ly-3, T-cell surface glycoprotein, T-cell surface glycoprotein CD8 alpha chain, T-cell surface glycoprotein CD8 beta chain, T-cell surface glycoprotein Lyt-2, T-cell surface glycoprotein Lyt-3, T-cell surface molecule, T-lymphocyte differentiation antigen T8/Leu-2, type I transmembrane glycoprotein | |
| IgG2a κ | |
| Affinity chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title