missing translation for 'onlineSavingsMsg'
Learn More

CDDO Methyl Ester, TRC

Product Code. 30355323
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
100mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30355323 5 mg 5mg
30354044 100 mg 100mg
30359091 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30355323 Supplier Toronto Research Chemicals Supplier No. TRCC2281005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 218600-53-4
Molecular Formula C32 H43 N O4
Molecular Weight (g/mol) 505.69
Synonym 2-Cyano-3,12-dioxooleana-1,9(11)-dien-28-oic Acid Methyl Ester, Bardoxolone Methyl, CDDO-Me, Methyl 2-cyano-3,12-dioxooleana-1,9(11)-dien-28-oate, NSC 713200, RTA 402
IUPAC Name methyl (4aS,6aR,6bS,8aR,12aS,14aR,14bS)-11-cyano-2,2,6a,6b,9,9,12a-heptamethyl-10,14-dioxo-1,3,4,5,6,7,8,8a,14a,14b-decahydropicene-4a-carboxylate
SMILES COC(=O)[C@]12CCC(C)(C)C[C@H]1[C@H]3C(=O)C=C4[C@@](C)(CC[C@H]5C(C)(C)C(=O)C(=C[C@]45C)C#N)[C@]3(C)CC2

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C32H43NO4/c1-27(2)11-13-32(26(36)37-8)14-12-31(7)24(20(32)17-27)21(34)15-23-29(5)16-19(18-33)25(35)28(3,4)22(29)9-10-30(23,31)6/h15-16,20,22,24H,9-14,17H2,1-8H3/t20-,22-,24-,29-,30+,31+,32-/m0/s1
Formula Weight 505.3192
Chemical Name or Material CDDO Methyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.