missing translation for 'onlineSavingsMsg'
Learn More

Eurofins DiscoverX CDK4/Cyclin D3 Protein, Active

Product Code. 17340854
Encompass_Preferred
Click to view available options
Quantity:
10 μg
250 μg
Unit Size:
10µg
250µg
This item is not returnable. View return policy

Product Code. 17340854

Brand: Eurofins DiscoverX 14957

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

N-terminal GST-tagged, recombinant, human CDK4 full length and N-terminal GST-tagged, recombinant, human cyclin D3 full length co-expressed by baculovirus in Sf21 insect cells. for use in Kinase Assays

CDK4 (Cyclin-dependent kinase 4) forms a complex with type D cyclins such as D1, D2 and D3 and when the cellular levels of cyclin D peak there is a shortening of the G1 phase and rapid entry into S phase of cell cycle progression. The substrate for cdk4/cyclin D is Rb, and phosphorylation of Rb results in dissociation of the E2F complex from Rb. The free E2F proteins in turn stimulate transcription of genes required for DNA synthesis. Cellular inhibition of CDK4/cyclin D3 activity is achieved by its inhibitory proteins, i.e. p16, p21 and p27. Inhibition of CDK4/CCND3 may be used in treatment of uncontrolled cell proliferation, tumor heterogeneity, invasion and metastasis.

GenBank NM_000075.3 / GenBank M90814.1

Specifications

Formulation mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% β-mercaptoethanol. Frozen solution.
Molecular Weight (g/mol) 61 kDa (CDK4) and 59 kDa (Cyclin D3)
Quantity 10 μg
Species Human
Recombinant Recombinant
Protein Tag N-terminal GST / N-terminal GST
Expression System Recombinant enzyme expressed in Sf21 insect cells
Protein Form Full-length
Purity or Quality Grade Combined ≥72% ratio 2-3:1 CDK4:cyclinD3
Protein CDK4/Cyclin D3, active
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.