missing translation for 'onlineSavingsMsg'
Learn More

Vileda Professional™ CE Duo™ CleanTech™ Sterile Sponge

Product Code. 17812496
Change view
Click to view available options
Autoclavable:
Autoclavable
Unit Size:
Pack of 60
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Autoclavable Unit Size
17812496 Autoclavable Pack of 60
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17812496 Supplier Vileda Professional™ Supplier No. 161622
  • ONLINE EXCLUSIVE PRICE
    853.00 €
    734.91 € / Pack of 60
    Save 118.09 €
    13% Off
Estimated Shipment: 29-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CE Duo™ Mopping System is an efficient and reliable cleaning system for contamination control. CE Duo™ CleanTech™ Sponge allows users to disinfect while cleaning hard-to-reach areas in cleanrooms and controlled environments.

  • Disinfect hard to reach areas in cleanrooms
  • Specially formulated microfiber foam provides a high degree of liquid release to achieve contact times
  • Excellent cleaning results, 3rd party proven 99.997% bacteria reduction
  • Based on special absorbency and lay down properties, this sponge offers valuable cost-in-use benefits specific to time and disinfectant use
  • Manufactured and sterilized in the USA
  • Validated to 10-6 SAL per ISO 11137
  • Individually double bagged with Lot # and Indicator Dot on each package
  • Certificate of Sterility available upon request with each shipment

Specifications

Product Type Cleanroom Sponge
Color Blue, White
Sterility Sterile
Autoclavable Autoclavable
Size 4 x 7 in.
Disposable Single-use
Quantity 6/Pk., 10 Pk./Cs.
Description Microfiber Cleaning and Disinfecting for Hard to Reach Areas
For Use With (Application) Cleaning and disinfecting behind equipment, piping, door handles, hoods, isolators, cabinets and any surfaces a mop can not reach.
Certifications/Compliance Certificate of Sterility upon Request
Color-coded No
Material Microfiber
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.