missing translation for 'onlineSavingsMsg'
Learn More

Cebranopadol, TRC

Product Code. 30368353
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30368353 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30368353 Supplier Toronto Research Chemicals Supplier No. TRCC22881010MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cebranopadol
CAS 863513-91-1
Molecular Formula C24H27FN2O
Quantity 10 mg
Synonym trans-6'-Fluoro-4',9'-dihydro-N,N-dimethyl-4-phenyl-spiro[cyclohexane-1,1'(3'H)-pyrano[3,4-b]indol]-4-amine, (1alpha,4beta)-6'-Fluoro-4',9'-dihydro-N,N-dimethyl-4-phenylspiro[cyclohexane-1,1'(3'H)-pyrano[3,4-b]indol]-4-amine, GRT 6005
InChI Formula InChI=1S/C24H27FN2O/c1-27(2)23(17-6-4-3-5-7-17)11-13-24(14-12-23)22-19(10-15-28-24)20-16-18(25)8-9-21(20)26-22/h3-9,16,26H,10-15H2,1-2H3
SMILES CN(C)[C@@]1(CC[C@]2(CC1)OCCc3c2[nH]c4ccc(F)cc34)c5ccccc5
IUPAC Name 6-fluoro-N,N-dimethyl-1'-phenylspiro[4,9-dihydro-3H-pyrano[3,4-b]indole-1,4'-cyclohexane]-1'-amine
Molecular Weight (g/mol) 378.48
Formula Weight 378.2107
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.