missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cecropin A (1-7) -Melittin (2-9) Hybrid Peptide

Product Code. 15408219
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
Unit Size:
0.5mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15408219 1 mg 1mg
15490637 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15408219 Supplier Thermo Scientific Chemicals Supplier No. J66741.M^
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

For cellular and molecular biology applications

Causes the release of carboxyfluoresceine encapsulated in phosphatidylcholine liposomes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Purification Method HPLC
Common Name [CA(1-7)M(2-9)NH2]
Content And Storage -30°C to -10°C
Format Lyophilized Powder
For Use With (Application) Causes the release of carboxyfluoresceine encapsulated in phosphatidylcholine liposomes.
Molecular Weight (g/mol) 1770.29
Product Type Cecropin A (1-7) -Melittin (2-9) Hybrid Peptide
Quantity 1 mg
Regulatory Status RUO
Sequence Lys-Trp-Lys-Leu-Phe-Lys-Lys-Ile-Gly-Ala-Val-Leu-Lys-Val-Leu-NH2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.