missing translation for 'onlineSavingsMsg'
Learn More

Cedrol

Product Code. 16621822 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16621822 100 g 100g
16601822 5 g 5g
16611822 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16621822 Supplier Thermo Scientific Acros Supplier No. 461421000
 Restricted Item
Estimated Shipment: 01-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

222.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Molecular Weight (g/mol) 222.37
Product Type Cedrol
Quantity 100 g
Solubility Information Solubility aq. soln.: insoluble. Other solubilities: soluble in chloroform

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.