missing translation for 'onlineSavingsMsg'
Learn More

Cefixime, TRC

Product Code. 30134709
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134709 1 g 1g
30137216 10 mg 10mg
17932795 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134709 Supplier Toronto Research Chemicals Supplier No. TRCC2428001G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79350-37-1
Molecular Formula C16 H15 N5 O7 S2
Molecular Weight (g/mol) 453.45
Synonym (6R,7R)-7-[[(2Z)-2-(2-amino-4-thiazolyl)-2-[(carboxymethoxy)imino]acetyl]amino]-3-ethenyl-8-oxo-5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[(2-amino-4-thiazolyl)[(carboxymethoxy)imino]acetyl]amino]-3-ethenyl-8-oxo-, [6R-[6α,7β(Z)]]-, 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[(2Z)-(2-amino-4-thiazolyl)[(carboxymethoxy)imino]acetyl]amino]-3-ethenyl-8-oxo-, (6R,7R)- (9CI), (-)-Cefixim, CL 284635, Cefexy, Cefixim, Cefixime, Cefiximine, Cefixoral, Cefoceb, Cefspan, Ceftas, Cephoral, FK 027, FR 17027, Fixitil, Hifen, Loprax, Oroken, Suprax, Suprax (antibiotic), Taxim-O, Unixime, Ximacef
IUPAC Name (6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(carboxymethoxyimino)acetyl]amino]-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
SMILES Nc1nc(cs1)\C(=N\OCC(=O)O)\C(=O)N[C@H]2[C@H]3SCC(=C(N3C2=O)C(=O)O)C=C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C16H15N5O7S2/c1-2-6-4-29-14-10(13(25)21(14)11(6)15(26)27)19-12(24)9(20-28-3-8(22)23)7-5-30-16(17)18-7/h2,5,10,14H,1,3-4H2,(H2,17,18)(H,19,24)(H,22,23)(H,26,27)/b20-9-/t10-,14-/m1/s1
Formula Weight 453.0413
Percent Purity >95%
Chemical Name or Material Cefixime
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.