missing translation for 'onlineSavingsMsg'
Learn More

Cefodizime Sodium, TRC

Product Code. 30355785
Change view
Click to view available options
Quantity:
1.5 g
3 g
Unit Size:
1.5g
3g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30355785 1.5 g 1.5g
30356745 3 g 3g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30355785 Supplier Toronto Research Chemicals Supplier No. TRCC2428831.5G
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86329-79-5
Molecular Formula C20H18N6Na2O7S4
Molecular Weight (g/mol) 628.63
Synonym Cefodizime Disodium, Cefodizime Disodium Salt, HR 221, Kenicef, S 77-1221B, THR 221, Timecef, (6R,7R)-7-[[(2Z)-(2-Amino-4-thiazolyl)(methoxyimino)acetyl]amino]-3-[[[5-(carboxymethyl)-4-methyl-2-thiazolyl]thio]methyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid Disodium Salt, [6R-[6alpha,7beta(Z)]]-7-[[(2-Amino-4-thiazolyl)(methoxyimino)acetyl]amino]-3-[[[5-(carboxymethyl)-4-methyl-2-thiazolyl]thio]methyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid Disodium Salt
IUPAC Name disodium;(6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-3-[[5-(carboxylatomethyl)-4-methyl-1,3-thiazol-2-yl]sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
SMILES [Na+].[Na+].CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(=C(N2C1=O)C(=O)[O-])CSc3nc(C)c(CC(=O)[O-])s3)\c4csc(N)n4

Specifications

Quantity 1.5 g
InChI Formula InChI=1S/C20H20N6O7S4.2Na/c1-7-10(3-11(27)28)37-20(22-7)36-5-8-4-34-17-13(16(30)26(17)14(8)18(31)32)24-15(29)12(25-33-2)9-6-35-19(21)23-9;;/h6,13,17H,3-5H2,1-2H3,(H2,21,23)(H,24,29)(H,27,28)(H,31,32);;/q;2*+1/p-2/b25-12-;;/t13-,17-;;/m1../s1
Formula Weight 627.9915
Chemical Name or Material Cefodizime Sodium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.