missing translation for 'onlineSavingsMsg'
Learn More

Cefpodoxime Combination Disc Kit

Product Code. 11912922
Change view
Click to view available options
Quantity:
50 Discs/Cartridge (250 Discs/Pk.)
Unit Size:
250 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11912922 50 Discs/Cartridge (250 Discs/Pk.) 250 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11912922 Supplier Thermo Scientific Oxoid Supplier No. DD0029B
 Restricted Item
Estimated Shipment: 04-01-2027
Añadir a la cesta
Añadir a la cesta

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

Detect ESBLs (extended spectrum β-lactamases) in Enterobacteriaceae which don't produce inducible AmpC enzymes with these diagnostic discs.

Detect ESBLs (extended spectrum β-lactamases) in Enterobacteriaceae which don t produce inducible AmpC enzymes with Thermo Scientific™ Oxoid™ Cefpodoxime Combination Disc Kit. The test combines the use of two discs: A disc with both Cefpodoxime and clavulanic acid (ß-lactamase inhibitor) and a disc with Cefpodoxime alone.

Extended Spectrum β-Lactamases are enzymes capable of conferring bacterial resistance to penicillins, cephalosporins, and aztreonam by hydrolysis of these antibiotics1,2. Production of ESBLs is a significant resistance-mechanism that impedes the antimicrobial treatment of infections caused by Enterobacteriaceae and is a serious threat to the currently available antibiotic armory3.

Use Cefpodoxime Combination Disc Kit as a rapid and simple method for the detection of ESBLs in Enterobacteriaceae.
  • Rapid Results
  • Simple and ready-to-use cartridge format.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY

Especificaciones

Content And Storage -20°C to +8°C
Description Cefpodoxime Combination Disc Kit
Concentration CD01: Cefpodoxime/clavualanic acid 10/1μg;  CPD10: Cefpodoxime 10μg
Detectable Analytes Extended spectrum beta lactamases (ESBLs)
Type Cefpodoxime Combination Disk Kit
Packaging Type 5 Cart./Pk.
Quantity 50 Discs/Cartridge (250 Discs/Pk.)
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.